Corning® Preassembled Closed Systems Solution Centrifuge Tubes, with Dip Tube, 50 ml, Corning
Supplier: Corning
11705
89214-346CS 171.56 USD
89214-346
Corning® Preassembled Closed Systems Solution Centrifuge Tubes, with Dip Tube, 50 ml, Corning
Tubes Centrifuge Tubes
Corning® 50 ml clear polypropylene (PP) preassembled closed systems self-standing centrifuge tubes.
  • Advanced polyethylene CentriStar™ caps
  • Conical self-standing bottom style, with graduations and diptube
The CentriStar™ cap has an easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping. This leak-proof design comes with a revolutionary plug feature that virtually eliminated all seepage when used under recommended conditions.
Please check our Technical Information Listing forMore Product Information