SPE Vacuum Manifolds, Bonna-Agela Technologies
Supplier: Phenomenex
VM12 VM24
89213-858EA 3938.1 USD
89213-858 89213-860
SPE Vacuum Manifolds, Bonna-Agela Technologies
Manifolds
SPE vacuum manifolds are ideal for SPE sample preparation, filtration and elution.
  • Provides consistent extraction and filtration results
  • Solvent resistant and autoclavable lid
  • Adjustable racks will accommodate a variety of sample collection vessels