Venusil® SAX HPLC Columns, Bonna-Agela Technologies
Supplier: Phenomenex
VSA950505-0
89213-442EA 1023.54 USD
89213-442
Venusil® SAX HPLC Columns, Bonna-Agela Technologies
Chromatography Columns
Bonded with quaternary amine group.
Good choice for basic compounds, water soluble polar compounds and bio-molecular.