Envoy® Diagnostic Ready Engineered Shippers, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
ERWS2LE472 ERWS2LE473 ERWS2LE570 ERWS2LEIN2
89213-117CS 992.34 USD
89213-117 89213-119 89213-125 89213-127
Envoy® Diagnostic Ready Engineered Shippers, Sonoco ThermoSafe
Mailers and Shippers
Envoy Diagnostic is a 2 to 8°C insulated shipping system that has been design-tested to ISCsilver™ ambient temperature profile to address the need for temperature assurance in moderate shipping conditions.
  • Convenient all-in-one solution with proven thermal performance
  • Ideal for 1- to 2-day shipments and small payloads
  • Packouts incorporate foam bricks for uniform temperature coverage
A simple, economical solution for refrigerated temperature control of diagnostic kits, blood samples, and other diagnostic specimens.