Digital 'Suction Type' Refractometers, ATAGO®
Supplier: Atago
3353
89212-808EA 2744.16 USD
89212-808
Digital 'Suction Type' Refractometers, ATAGO®
Refractometers Handheld Refractometers
Model QR-BRIX, vaccum is created and released to draw the liquid inside
The device features IP64-rated dustproof and water-resistant protection, and Automatic Temperature Compensation for accurate measurements in shifting environmental conditions. A sample volume of 2mL is required for measurement results.
Cleaning process is easy and prism chamber can be opened to remove thr residues