Lovibond® RD 125 Reactor
Supplier: Lovibond
2418940
89212-744EA 1778.45 USD
89212-744
Lovibond® RD 125 Reactor
Block Heaters Digital Block Heaters
The RD 125 thermoreactor can be used for the tube test digestion of highly sensitive and precise COD (150 °C), TOC (120 °C), total chromium (100 °C), total nitrogen (100 °C) and total phosphate (100 °C) determinations.
  • With the voltage switch on the back either 220 and 110 V can be chosen
The required temperatures and reaction times are pre-programmed, and easy to select by using the keypad. Users can pre-select reaction times of 30, 60 and 120 minutes or can set it to operate continuously. When digestion is complete, the reactor automatically turns off and will sound an alarm.
The RD 125 Reactor is fitted with 24 holes for 16 mm diameter vials.