MD 500 Multiparameter Photometer, Lovibond®

Supplier: Lovibond
214020
89212-658EA 2495.8 USD
89212-658
MD 500 Multiparameter Photometer, Lovibond®
Colorimeters Multiparameter Colorimeters
The MD 500 is a fully portable 6 LED, multiparameter photometer. MD 600 is a completely functional integrated unit designed for the outdoor professional for on-site visits.

  • Contains more than 120 programmed methods
  • Waterproof, easy to handle
  • Automatic wavelength selection
  • Uses tablet, liquids, powders or vials
  • Storage of up to 1000 data sets

The modern design of the MD 500 succeeds in combining the mobility of a portable photometer with the characteristics of a modern laboratory photometer

The MD 500 can also be used with the optional infrared data transfer module which is supplied complete with data logging software, providing additional data manipulation tools and allowing the data to be saved as a spreadsheet or text file.

This unit covers all the important parameters of water analysis, from aluminium to zinc. The high level of accuracy of Lovibond® reagents and the user-friendly nature of the instrument guarantee rapid and reliable analysis.

Ordering information: The pack includes with carrying case, 4×AA/LR6 batteries, 3× round glass vials 13 mm, 3× round glass vials 16 mm,1 adapter each for 16 mm and 13 mm vials, plastic stirring rod, brush, screw driver, guaratee sheet, certificate of compliance and instruction manual.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR