Mini Safety Cabinet, Justrite®
Supplier: Justrite
890200
89209-854EA 730.94 USD
89209-854
Mini Safety Cabinet, Justrite®
Furniture Storage Cabinets
This safety cabinet offers the smallest footprint for storing small quantities of flammables for cleaning and maintenance at point-of-use applications
  • Color: yellow
The cabinet includes three bungee cords to keep contents in place when transporting cabinet on service vehicles, carts or maintenance trikes.