Inoculation Loops, 1 µl, with Calibration Certificate, Globe Scientific
Supplier: Globe Scientific
2821
89209-784CS 180.67 USD
89209-784
Inoculation Loops, 1 µl, with Calibration Certificate, Globe Scientific
Spreaders Inoculating Loops and Needles
These 1uL disposable inoculation loops offer a convenient and safe way of streaking culture media
These loops are enclosed in peel-apart packages, with each pack containing 20 loops. All loops are sterilized by gamma radiation.
Molded from a special grade of polystyrene (PS), they offer a strong, rigid feel to provide comfortable, stable and uniform streaking.
Certifications: Each case comes with a Certificate of Calibration.