20 L Jacketed Filter Reactor System, Electric Motor, Chemgalss
Supplier: Chemglass
CG-1959-F620
89209-354EA 68099.39 USD
89209-354
20 L Jacketed Filter Reactor System, Electric Motor, Chemgalss
Laboratory Reactor Systems
Jacketed filter glass reactor features single or multi-step.
  • Complete reactor system on open air, autocenter stand
  • 150 °C maximum temperature limit for filter base
  • Filter base eliminates material transfers and significantly reduces losses
Can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis.