Jacketed Filter Reactor Systems, 20 L, Air Motor, Chemglass
Supplier: Chemglass
CG-1958-F621
89209-344EA 61105.7 USD
89209-344
Jacketed Filter Reactor Systems, 20 L, Air Motor, Chemglass
Laboratory Reactor Systems
Jacketed filter glass reactors feature autocenter stand.
  • Complete reactor system on open air, autocenter stand
  • 150 °C maximum temperature limit for filter base
  • Filter base eliminates material transfers and significantly reduces losses
Jacketed filter glass reactors can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis.