10 L Jacketed Filter Reactor Systems, Air Motor, Chemglass
Supplier: Chemglass
CG-1958-F611
89209-342EA 58919.14 USD
89209-342
10 L Jacketed Filter Reactor Systems, Air Motor, Chemglass
Laboratory Reactor Systems
Complete reactor system on open air, autocenter stand.
Maximum temperature limit 150 °C for filter base.