Stainless Steel Cleaner, Micronova Manufacturing Inc.
Supplier: Micronova
SC1-P
89208-418CS 158.68 USD
89208-418
Stainless Steel Cleaner, Micronova Manufacturing Inc.
Cleaning Agents Detergents and Surface Cleansers
Gel Stainless Steel Cleaner uses advanced no-drip technology to eliminate streaks and drips when applied to stainless, metal and other hard surfaces.
Spray gel clings on contact and glides over most surfaces with minimum effort. When dealing with rust, oxidation and residue build up, the gel can be applied to seams and crevices to pre-treat before wiping.
The gel is not atomized into ambient air and so will not impact indoor air quality. Contains no phosphorus and has biodegradable surfactants. No VOC's.