SHOWA Atlas 380 Foam Nitrile Palm Coated Glove, Showa
Supplier: Showa
380M-07 380XL-09 380S-06 380L-08
89208-330CS 2121.73 USD
89208-330 89208-334 89208-328 89208-332
SHOWA Atlas 380 Foam Nitrile Palm Coated Glove, Showa
Gloves
Ideal glove for applications requiring extreme grip, dexterity, and comfort.
  • Foam waffle pattern surface offers excellent grip
  • 13 gauge liner provides flexibility
  • Extended cuff for added protection
Engineered with a lightweight, 13 gauge seamless knit liner and waffle-pattern foam nitrile palm coating to provides optimal grip across a wide range of applications.
Ordering information: Order by Master case of 72 pair.
Packaging: Packaged 12 pair per poly bag, 6 bags per master case.