Midstream Pregnancy Test, Clarity Diagnostics
Supplier: Clarity Diagnostics
OTC-HCGMID2
89207-592CS
566.46
USD
89207-592
Midstream Pregnancy Test, Clarity Diagnostics
Clinical Diagnostic Supplies
Clinical Reagents and Kits
This HCG midstream test is OTC approved for home use.
- With QSG for ease of use
- Detects pregnancy first day of missed period
- Results within 1 to 5 minutes
- Sensitivity 25 mIU/ml
- Can be stored at room temperature
- 16 to 18 month dating
Certifications: FDA Cleared and OTC Approved.
Packaging: Individually packed in foil pouch. Convenient and affordable two tests/pack.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)