Q55 Sonicator, QSonica, LLC.
Supplier: QSonica, LLC.
Q55-110
89207-044EA 4051.22 USD
89207-044
Q55 Sonicator, QSonica, LLC.
Homogenizers Ultrasonic Cell Disruptors
Q55 sonicator is a compact ultrasonic processor occupies less bench space effective for standard cell disruption and many other small volume applications.
  • With three different sizes probes
  • Simple and effective operation
  • Thumb-switch or continuous operation
Ordering information: It includes generator, converter, diameter probe (3 mm), power cable, converter cable and wrench set and the stand must be ordered separately.