Orion™ Versa Star™ pH/ISE Measurement Module, Thermo Scientific
Supplier: Thermo Fisher Scientific
VSTAR-ISE
89206-420EA 1483.32 USD
89206-420
Orion™ Versa Star™ pH/ISE Measurement Module, Thermo Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Add pH, ion concentration, mV, ORP, and temperature measurement capabilities to a Versa Star™ meter using the Orion™ Versa Star™ pH/ISE Measurement Module.
  • Measure pH, ion concentration, mV, relative mV, or ORP with temperature
  • Advanced ISE features include non-linear selectable auto-blank, low range stability, and electrode ID with calibration verification
  • 10 methods per channel with password protection
  • Three-year module warranty
Add pH, ion concentration, mV, ORP, and temperature measurement capabilities to an Orion™ Versa Star™ meter using the Orion™ Versa Star™ pH/ISE Measurement Module
Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN, or user-defined buffer groups with choice to manually enter custom buffer values. Perform up to a six point ISE (Ion Selective Electrode) calibration with selectable units of ppm, M, mg/L, %, ppb, or none. Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH or ion concentration reading.
Each Orion™ Versa Star™ meter has four channels that can be customized with interchangeable measurement modules. The Versa Star™ pH/ISE module offers up to six point pH and ISE calibrations and advanced ISE features, including incremental techniques (single and double known addition and subtraction, analate addition and subtraction), low range stability, and electrode ID with calibration verification.