Orion™ Versa Star™ pH Measurement Module, Thermo Scientific
Supplier: Thermo Fisher Scientific
VSTAR-PH
89206-414EA 784.18 USD
89206-414
Orion™ Versa Star™ pH Measurement Module, Thermo Scientific
Electrochemistry Meters pH and Multi-Parameter Meters
Add pH, mV, ORP, and temperature measurement capabilities to Versa Star™ meters using the Orion™ Versa Star™ pH Measurement Module.
  • Measure pH, mV, relative mV, or ORP with temperature
  • Perform up to a six point pH calibration using automatic buffer recognition of NIST, DIN, or user-defined buffer groups
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Customize measurements to show calibration history and pH electrode condition or simplify to show an extra large pH reading
  • 10 methods per channel with password protection
  • Three-year module warranty
Add pH, mV, ORP, and temperature measurement capabilities to an Orion™ Versa Star™ meter using the Orion™ Versa Star™ pH Measurement Module
Select from five Orion™ Versa Star™ measurement modules to customize the measurement capabilities of an Orion™ Versa Star™ meter. Measurement modules are interchangeable and can be added or removed from each Versa Star™ meter's four channels at any time.
Each Versa Star™ meter has four channels that can be customized with interchangeable measurement modules. The Versa Star™ pH module offers up to a six point pH calibration with easy to follow onscreen instructions, automatic buffer recognition of NIST, DIN, or user-defined buffer groups, and calibration editing option to fix individual points without a full recalibration.