PIG® Plug-N-Seal® Patching Paste, New Pig
Supplier: New Pig
PTY220
89205-886EA 37.02 USD
89205-886
PIG® Plug-N-Seal® Patching Paste, New Pig
Spill Control Materials
Temporarily stops leaks until system can be drained or leak source contained
No preparation needed, even on dirty or rusty surfaces. Temporarily stops leaks from 2 minutes up to 2 hours. Ready to use without mixing.
Excellent chemical resistance makes it excellent for flammable or hazardous leaks.