Inland 45 Pump Oil, Restek
Supplier: Restek
24819
89205-674EA 157.52 USD
89205-674
Inland 45 Pump Oil, Restek
Lubricants
Recommended for optimum mass spec performance, this oil pump is compatible with buna-N, neoprene, and Viton® seals.
  • Nontoxic
  • Noncorrosive
  • Ease at cold start
  • Optimum vacuum pump performance
This pump oil functions at low vapor pressure 10-7 torr.