Corning® 2 L Polycarbonate Erlenmeyer Flask, Corning
Supplier: Corning
431518
89204-710CS 1415.69 USD
89204-710
Corning® 2 L Polycarbonate Erlenmeyer Flask, Corning
Flasks Erlenmeyer Flasks
Corning Erlenmeyer Flasks with graduations are now available as closed system solutions.
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Flat bottom design without baffles
  • Supplied individually doubled-bagged and preassembled
  • Available for 1L, 2L and 3L flask sizes
The preassembled transfer caps have two ports. One port ends in a 0.2 μm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in either a male Luer lock or a male polycarbonate quick connect. The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes. Made from optically clear polycarbonate.