Corning® Erlenmeyer Flask, 2 L, Male Luer Lock, Corning
Supplier: Corning
431512
89204-704CS 1290.26 USD
89204-704
Corning® Erlenmeyer Flask, 2 L, Male Luer Lock, Corning
Flasks Erlenmeyer Flasks
Corning Erlenmeyer Flasks are available as closed system solutions. They are sterile and arrive individually doubled-bagged and preassembled.
  • Flat bottom
  • Preassembled transfer cap
The preassembled transfer caps have two ports: One port ends in a 0.2 μm filter, and the other port is chemically resistant, heat sealable flexible tubing, ending in either a male Luer lock or a male polycarbonate quick connect
Corning Erlenmeyers are autoclavable once at 121°C, 15 psi for 20 minutes.
The dip tube reaches all the way to the bottom of the flask for easy aseptic transfer of your liquid handling processes.
Certifications: Certified non-pyrogenic and Rnase-/Dnase-free, and sterile.