Anti-Static Polyimide Cleanroom Tape with Silicone Adhesive, UltraTape
Supplier: Ultratape Industries
0157BZ200P3S
89203-686CS 3024.95 USD
89203-686
Anti-Static Polyimide Cleanroom Tape with Silicone Adhesive, UltraTape
Tapes
Particle-free, residue-free tapes.
  • Thin and comformable
  • Puncture-, tear-, and chemical-resistant
  • Suitable for use in high temperatures
A Polyimide based film coated with an anti-static coating and silicone adhesive. Ideal for masking of printed circuit boards during wave soldering or hot air leveling processes.