LE621 Dissolved Oxygen Sensors with Temperature Probe, METTLER TOLEDO®
Supplier: Mettler Toledo
51340403
89203-172EA 585 USD
89203-172
LE621 Dissolved Oxygen Sensors with Temperature Probe, METTLER TOLEDO®
Electrodes Dissolved Oxygen Electrodes
LE621 galvanic dissolved oxygen electrode with integrated temperature sensor (NTC 30 KΩ), designed for use with METTLER TOLEDO® DO meters (FG4 only)
  • Galvanic
  • Integrated temperature sensor
  • BNC/Cinch connector
  • PVC shaft
Features BNC/Cinch connector and PVC shaft.