VWR® Angled Thermometer Storage Trays
Supplier: VWR International
61400-5111
89203-047PK 112.55 USD
89203-047
VWR® Angled Thermometer Storage Trays
Trays/Dishes/Pots Instrument Trays/Dishes
Recyclable trays feature a built-in angle to prevent separation of thermometer liquid.
  • UV coated
  • Made from 2mm polystyrene
3- or 14-slot versions available with numbered slots.