VWR® Immersion Circulators
Supplier: VWR International
89202-886
89202-886EA 1207.6 USD
89202-886
VWR® Immersion Circulators
Circulating Thermostats Open Circulator Systems Thermostat Controllers
Create your own circulating bath system using your own vessel or a VWR® open bath tank.
  • User-adjustable temperature limits and alarms
  • Convenient slide control
  • Single-point calibration
Large, 8.3 cm (31/4") universal icon and display. User-adjustable high- and low-temperature limits and alarms, as well as integral pump and heater coil protection. Convenient slide control allows easy adjustments of flow rate. Single-point calibration.
Certifications: ETL listed. CE marked. Complies with DIN 12876-1 Class I safety requirements for use with non-flammable liquids. cETLus compliant.
Ordering information: 240V/50Hz models are also available; contact your Avantor sales representative for more information.