Digital MicroPlate Genie® Microplate Mixer Shaker, 230 V
Supplier: Scientific Industries
SI-0401A
89202-332EA 1431.31 USD
89202-332
Digital MicroPlate Genie® Microplate Mixer Shaker, 230 V
Shakers and Mixers Microplate Shakers
Fully digital programmability for precise speed and time control and a pulsing feature are incorporated in this mixer which has an aggressive speed for increased mixing efficiency in every well, even 384-well plates.
  • 230 V model
  • Fully programmable
  • Precise aggressive speed
  • Pulsing feature
The mixers high speed and small orbit combine to offer true vortexing for complete and uniform mixing. A built-in digital timer operates continuously or from 0 to 99 minutes.
Certifications: CE marked.
Ordering information: The 230 V model is supplied without a plug.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (89202-330) is also available, please see separate product page.