Digital MicroPlate Genie® Microplate Mixer Shaker, Scientific Industries
Supplier: Scientific Industries
SI-4010 SI-0400A
87004-336EA 172.73 USD
87004-336 89202-330
Digital MicroPlate Genie® Microplate Mixer Shaker, Scientific Industries
Shakers and Mixers Microplate Shakers
Fully digital programmability for precise speed and time control and a pulsing feature are incorporated in this mixer which has an aggressive speed for increased mixing efficiency in every well, even 384-well plates.
  • Fully programmable
  • Precise aggressive speed
  • Pulsing feature
The mixers high speed and small orbit combine to offer true vortexing for complete and uniform mixing. A built-in digital timer operates continuously or from 0 to 99 minutes.
Certifications: CE marked.
Ordering information: The 230V model is supplied without a plug.