QuadMag Genie® Four-Position Magnetic Stirrers, Scientific Industries
Supplier: Scientific Industries
SI-5236
89202-326EA 1579.77 USD
89202-326
QuadMag Genie® Four-Position Magnetic Stirrers, Scientific Industries
Stirrers Magnetic Stirrers
These four-position magnetic stirrers are available in an analog model or digital model
Choose the analog model, which is optimized for high speed, or choose the digital model for better speed control; this model offers digital display, control of speed, and is time-speed adjustable in 10rpm increments. Digital models also incorporate a timer from 1 to 99 minutes and a ramping feature that slowly increases speed to avoid splashing and improve spin bar control. Can be used in cold rooms and incubators.
Powerful, general purpose four-position magnetic stirrers feature "rare earth" magnets that stay coupled under the most demanding strirring conditions and offer exceptional magnetic force capable of stirring up to 2L for each position
Certifications: CE marked.