CentriVap® Cold Traps, –105 °C, Labconco
Supplier: Labconco
7385020 7385030 7385035 7385037 7385040 8621170 8621190 8621100 8621110 8621115 8621130 8621140 8621150
89202-060EA 10589.16 USD
89202-060 89202-062 89202-064 89202-066 89202-068 78008-018 78008-020 78008-006 78008-008 78008-010 78008-012 78008-014 78008-016
CentriVap® Cold Traps, –105 °C, Labconco
Vacuum Traps
Cold traps collect evaporated liquid from the CentriVap® Vacuum Concentrator, protecting the vacuum pump from corrosion
These traps may also be used in other laboratory applications such as freeze drying and desiccation.
Ordering information: The CentriVap® cold trap is available separately or as a component of CentriVap® concentration systems and CentriVap® gel dryer systems. Additional available CentriVap® cold traps include –50 °C and –84 °C models; contact your Avantor representative for more information.