Professional’s Choice® Steril-Sure® Self-Seal Pouches, GS Medical Packaging
Supplier: GS Medical Packaging
442089
89201-581CS 370.07 USD
89201-581
Professional’s Choice® Steril-Sure® Self-Seal Pouches, GS Medical Packaging
Sterilization Wraps
Self-seal pouches are suitable for sterilizing articles in any healthcare or personal services settings where sterilization and infection control practices are required.
  • Lead-free and medically safe inks
  • Latex-free pouches
Pouches are constructed of the finest surgical grade paper together with a multi-layer fracture-free tinted blue film to provide a clean, fibrefree peel. The self-adhesive tape has been tested in all sterilization conditions to ensure a 100% fail-resistant seal.
Certifications: Professional’s Choice® steam sterilization ink has been tested and validated by an independent laboratory to prove that all three parameters required for effective steam sterilization (time, temperature and steam) have been met. The ink has been verified to be in compliance with ISO 11140-1:2005.