STERIL-PEEL™ Heat Seal Pouches, GS Medical Packaging
Supplier: GS Medical Packaging
221633
89201-554CS 687.07 USD
89201-554
STERIL-PEEL™ Heat Seal Pouches, GS Medical Packaging
Sterilization Indicators
All STERIL-PEEL™ products are manufactured from medical grade bleached kraft paper
STERIL-PEEL™ products are printed with dual indicators which change color in either steam or ethylene oxide. The steam indicator ink will only change in the presence of all necessary parameters (saturated steam, temperature, and time).
Hydrogen peroxide, rather than chlorine, is used as the bleaching agent. They are made from virgin pulp, ensuring optimum consistency of the paper sheet, providing superior barrier properties.
Certifications: Both indicators conform with the latest ISO 11140 and EN868 regulations. 510K and CE compliant.