VWR® Disposable N95 Particulate Respirators
Supplier: VWR International
89201-508
89201-508CS 221.97 USD
89201-508
VWR® Disposable N95 Particulate Respirators
Respirators Disposable Respirators Particulate Respirators
VWR® N95 disposable respirators combine excellent safety and comfort features to ensure reliable protection and a lightweight, secure fit.
  • Meets NIOSH requirements for minimum filter efficiency of 95% against 0.3 µm particles
  • Provides protection against solid and liquid particles that do not contain oil
  • Lightweight construction allows for comfortable, extended use
An adjustable noseclip provides a secure seal against the user's face.