Forma™ Audible Temperature Monitors, Thermo Scientific
Supplier: Thermo Fisher Scientific
290003
89200-310EA 1566.4 USD
89200-310
Forma™ Audible Temperature Monitors, Thermo Scientific
Data Loggers
The compact monitor attaches to wall or equipment for convenient single-unit monitoring
  • Ringback system provides additional protection
  • Self-contained probe
  • Remote alarm contac
For use monitoring temperature of freezers, lab refrigerators, and incubators. Alerts to over- and under-temperature conditions or power loss, with alerm-reset feature. Battery back-up keeps monitor operational, even during a power failure.
Delivery information: Includes remote alarm contacts for connection to a Forma remote alarm.