Tissue-Tek Rapid Tissue Processors, Sakura
Supplier: Sakura Finetek
7720
89199-776EA 260265.39 USD
89199-776
Tissue-Tek Rapid Tissue Processors, Sakura
Tissue Processors
Continuous tissue processing utilizing low-wattage microwave technology, molecular-friendly reagents and vacuum infiltration for rapid turn around.
  • Throughput of up to 120 specimens per hour, loading 40 cassettes every 20 minutes (X120) and up to 45 specimens every 45 minutes (X50)
  • Biopsy and larger specimens can be processed in same run
2 USB ports, external alarm output, UPS signalinput, LAN connection
Ordering information: No formalin or xylene is used on the X120 processor.