Tissue-Tek® Accu-Edge® Pathology Workstation, Sakura Finetek
Supplier: Sakura Finetek
115
89199-666EA 27532.6 USD
89199-666
Tissue-Tek® Accu-Edge® Pathology Workstation, Sakura Finetek
Furniture Tables and Workbenches
The Tissue-Tek® Accu-Edge® Pathology Workstation is constructed of high-quality stainless steel and offers a comprehensive suite of standard and optional features.
  • Signed blueprints provided with signature required by facilities.
Standard Pathology Workstation features include: Dissecting board, Exhaust duct to building, Formalin container with spigot, Magnetic instrument holder, C-fold paper towel holder, Deck mounted magnifier and light, Dual recessed halogen lights, Standard sink, GFCI waterproof electrical receptacles, Vacuum breaker protection, Optional base legs (sold separately).
Accessories information: A wide assortment of accessory items and components designed for use with Tissue-Tek® Accu-Edge® Workstations and Grossing Stations.