Tissue-Tek® Accu-Edge® Countertop Grossing Station, Sakura Finetek
Supplier: Sakura Finetek
110
89199-658EA 25654.6 USD
89199-658
Tissue-Tek® Accu-Edge® Countertop Grossing Station, Sakura Finetek
Furniture Casework
Featuring enhanced grossing capabilities for limited spaces, the Tissue-Tek® Accu-Edge® Countertop Grossing Station is ideal for smaller labs, as it designed to sit on any countertop
  • Standard grossing station features include:
  • Adjustable leveling feet
  • Dissecting board
  • Two duct options: outside ventilation source or self-contained ventilation system with replaceable filters
  • Formalin container with spigot
  • GFIC waterproof electrical receptacles
  • Magnetic instrument holder
  • Deck mount magnifier light
  • C-fold paper towel holder
  • Dual recessed halogen lighting
  • Small sink
  • Vacuum breaker protection
  • Optional base legs (sold separately)
Signed blueprints provided with signature required by facilities.
Constructed from high-quality stainless steel, this countertop Grossing Station is equipped with a vacuum breaker-protected water supply.
Ordering information: For a detailed listed of optional accessory items and workstation components, see Tissue-Tek® Accu-Edge® Pathology Station Accessories, Sakura Finetek.