Feather A35 Low Profile Microtome Blades, Sakura
Supplier: Sakura Finetek
4980
89199-474CS 1768.82 USD
89199-474
Feather A35 Low Profile Microtome Blades, Sakura
Microtomes and Microtome Assemblies Microtome Blades and Handles
Low profile, ultra sharp blades section paraffin blocks without striations or chatter
Dispenser made to safely eject blades and store used blades. Blades are coated with a PTFE resin surface.
Ordering information: Blades are packaged with the dispenser. A case of 10 dispensers contains 50 blades per dispenser.