Paraffin wax blended with synthetic polymers, Solidification point: 56-57°C (132.8-134.6°F), pellets, Tissue-Tek® Paraform® processing/embedding medium Formula 3

Supplier: SAKURA FINETEK USA INC
Warning

7052
89199-380CS 167.86 USD
89199-380
Paraffin wax blended with synthetic polymers, Solidification point: 56-57°C (132.8-134.6°F), pellets, Tissue-Tek® Paraform® processing/embedding medium Formula 3
Paraffin wax blended with synthetic polymers

The unique composition of Tissue-Tek VIP® Paraffin allows for reduced compression sections as thin as 2 microns. The low melting point, 56°C, protects samples from heat damage. The paraffin will not discolor under normal processing conditions and leaves no plasticizer residue to clog paraffin lines in tissue processors and embedding centers. Tissue-Tek VIP Paraffin is supplied in small pellets for rapid melting.


  • For use with the infiltration and embedding of human or animal tissue to create paraffin blocks for routine and research histology
  • Unique paraffin composition
  • Low melting point
  • Small pellets
  • Compatible with automated embedding systems


Tissue-Tek Paraform Processing & Embedding Medium is a 3rd generation formulation designed for use with the Tissue-Tek AutoTEC® a120 Automated Embedding System for the fast, continuous and predictable embedding of blocks. The paraffin is designed as both a processing and embedding medium to cover all of the laboratory’s needs, with the same additional features as the Tissue-Tek VIP Paraffin.


Certifications: IVD, FDA Class I

CAS Number: 8002-74-2
IVDLogo

Order Now

Specification Test Results

Medium Composition Purified paraffin and synthetic resin blend
Applications Processing/Embedding
Additives None
Melting Point 56° to 58°C / 132° to 136°F
High Temperature Stability 65°C
Solid Product Range Pellets
Sectioning Range Down to 2µ


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR