VWR® Dual Centrifuge Tube Stand
Supplier: VWR International
89199-176
89199-176PK 126.8 USD
89199-176
VWR® Dual Centrifuge Tube Stand
Racks Tube Racks
The VWR® Dual Centrifuge Tube Stand is capable of holding both 15mL and 50mL centifuge tubes.
  • Stable base for funneling and filtering
  • Polyproylene construction
  • Low-cost alternative to metal centrifuge racks
This low-cost alternative to metal centrifuge racks provides a stable base for funneling and filtering. It also offers deep wells to allow users to read lower graduations. Manufactured from polypropylene.