Pharma-80 Tubing, Silicone, DuPont
Supplier: Liveo
3100651 4008432
89198-896PK 313.51 USD
89198-896 89198-898 89198-900 89198-894
Pharma-80 Tubing, Silicone, DuPont
Tubing Transfer Tubing
Pharmaceutical grade silicone tubing for transferring of ultra-pure liquids, air or steam in pharmaceutical and biotechnological manufacturing processes.
  • Excellent flexibility and low extractables
  • Easily sterilized
  • High resiliency
  • Complete traceability
  • No impaired taste or odor
Certifications: Meets or exceeds 21 CFR 177.2600, GMPs, USP Class VI and 3A Sanitary Standard procedures and European Pharmacopoeia Monograph 3.1.9. silicone elastomer for closures and tubing.
Delivery information: Standard package includes 15-meter coil, double bagged polyethylene bags in a cardboard box.