Low Speed Benchtop Centrifuge LMC-3000, Grant Instruments
Supplier: GRANT USA INC.
LMC-3000 USA
89194-510EA 3319 USD
89194-510
Low Speed Benchtop Centrifuge LMC-3000, Grant Instruments
Centrifuges Benchtop Centrifuges
The Grant Bio LMC-3000 is a low speed benchtop centrifuge designed for biomedical applications
Spin speed: up to 3000rpm for tubes, up to 2000rpm for microtitre plates
Timed operation (1 to 90 minutes), with automatic switch-off
‘Soft-start’ and ‘run-down’ of the rotor
Choice of interchangeable rotors for up to 12 centrifuge tubes or 2 microplates
Auto ‘IMBALANCE’ stop and warning diagnostics
Low noise level
Sturdy metal housing and lid, automatic imbalance switch-off and lid lock when centrifuge is running providing safe operation in your workplace
The centrifuge is suitable for a wide range of analytical applications including biomedical, bio-organic, and immunoenzyme analysis.
It features interchangeable rotors for accommodating centrifuge tubes (10 to 15mL, or 50mL) or standard 96-well microtitre plates.
Certifications: CE marked.
Ordering information: Units include a manufacturer's two-year warranty. Rotors must be ordered separately. Units are also available in a 230V format. Contact your Avantor representative for additional information.