VWR® Protective Eyewear with Adjustable Temples
Supplier: VWR International
89187-984
89187-984CS 1396.95 USD
89187-984
VWR® Protective Eyewear with Adjustable Temples
Safety Glasses
Ultra-lightweight nylon frame with soft PVC nose pads.
  • Unique ratcheting/adjustable temples lock into place for a custom fit
  • Ultra-lightweight nylon frames and soft PVC nose pads to prevent slipping
  • 8-base spherical lenses provide excellent side protection
  • Anti-fog coating extends lens life
Wrap-around sport styling, designed for men and women features eight base spherical lenses providing excellent side protection. The impact resistant polycarbonate lenses provide 99.9% UV protection and the scratch resistant coating.
Certifications: ANSI Z87.1