Spectrum Two™ FT-IR Spectrometer and Analysis Systems, PerkinElmer
Supplier: PerkinElmer
L160000F L1608010 L1608012 L1608004 L160000V L160000T L160000W L1600116 L1608005 L160000R L160000U L160000A L1608009 L1608007 L1608022 L160000Q L160000S L1600114 L1600129 L1600115 L1608006 L1608003 L1608013 L1608001
89187-894EA 39323.56 USD
89187-894 89187-918 89187-922 89187-940 89187-864 89187-866 89187-870 89187-916 89187-942 89187-882 89187-900 89187-908 89187-914 89187-924 89187-948 89187-878 89187-884 89187-910 89187-912 89187-920 89187-930 89187-938 89187-952 89187-966
Spectrum Two™ FT-IR Spectrometer and Analysis Systems, PerkinElmer
Spectrophotometers and Photometers IR and FTIR Spectrophotometers
A single software platform incorporates all of the functions required for infrared analyses, complete with instrument control, data manipulation and analysis, and flexible report utilities.
  • DynaScan interferometer
  • KBr optics
  • OpticsGuard™ technology
  • Atmospheric Vapor Compensation™ (AVC)
  • Compact design
  • Ready-made protocols
  • Materials library
  • Many resource packs for compliance with different industries
Spectrum Two FTIR Analysis Systems include the Spectrum Two instrument, Spectrum 10 or 10ES (Enhanced Security) software and the appropriate applicationMore Product Information