Spectrum Two™ FT-IR Spectrometer and Analysis Systems, PerkinElmer
Supplier: PerkinElmer
L160000F L1608010 L1608012 L1608004 L160000V L160000T L160000W L1600116 L1608005 L160000R L160000U L160000A L1608009 L1608007 L1608022 L160000Q L160000S L1600114 L1600129 L1600115 L1608006 L1608003 L1608013 L1608001
89187-894EA 39323.56 USD
89187-894 89187-918 89187-922 89187-940 89187-864 89187-866 89187-870 89187-916 89187-942 89187-882 89187-900 89187-908 89187-914 89187-924 89187-948 89187-878 89187-884 89187-910 89187-912 89187-920 89187-930 89187-938 89187-952 89187-966
Spectrum Two™ FT-IR Spectrometer and Analysis Systems, PerkinElmer
Spectrophotometers and Photometers IR and FTIR Spectrophotometers
A single software platform incorporates all of the functions required for infrared analyses, complete with instrument control, data manipulation and analysis, and flexible report utilities.
  • DynaScan interferometer
  • KBr optics
  • OpticsGuard™ technology
  • Atmospheric Vapor Compensation™ (AVC)
  • Compact design
  • Ready-made protocols
  • Materials library
  • Many resource packs for compliance with different industries
Spectrum Two FTIR Analysis Systems include the Spectrum Two instrument, Spectrum 10 or 10ES (Enhanced Security) software and the appropriate application pack
Academia: For teachers and students of spectroscopy in academia, Spectrum Two helps develop an understanding of IR sampling techniques and spectral analysis. The special educational resource pack facilitates teaching of traditional synthetic chemistry analysis as well as more modern applications, such as recycling.
Compact and robust, Spectrum Two is ideal for use in academic laboratories. Thanks to a unique power management system and OpticsGuard™ Spectrum Two can be switched in and out of standby without risk to sensitive optical components, significantly reducing power consumption and long term cost of ownership. Wireless operation and multi-user site licenses are also available for enhanced data system security in teaching environments.
Spectrum 10 and 10ES Software is fully validated and includes full instrument set-up and control functions, basic and advanced data manipulation routines, spectral arithmetic calculations with custom Equations Editor, COMPARE spectral comparison feature, PLS/PCR/Beer's Law predictions feature, Beer's Law Quant method development, Spectrum 10 easy macro writer, data tune-up, spectral searching, starter libraries, configurable reporting and data exporting, built-in tutorials and online help, laboratory scheduler feature, and Scanalyze for real-time results while scanning. Spectrum 10ES Software is 21 CFR Part 11 compliant, which encompasses access control, audit trailing, data security, and data integrity. It also includes report builders, report viewers, and administration utility.
Spectrometers and Application Packs can also be purchased individually.
Ordering information: Spectrum Two FTIR Analysis Systems include the Spectrum Two instrument, Spectrum 10 or 10ES software and the appropriate application pack. Instruments are supplied with mains cable, Ethernet and USB cables, electronic user's guide and maintenance guide on CD, certificate of conformity, and fuses. Each instrument includes a one-year manufacturer warranty and customer installation guide. Additional on-site warranties and installation are optional. Instrument purchases require the separate purchase of a power cord, suitable for 100-240V, 50/60 Hz operation.