Round Cover Glass, Azer Scientific
Supplier: Azer Scientific Inc.
200121
89187-782CS 666.26 USD
89187-782
Round Cover Glass, Azer Scientific
Cover Glasses
The round cover glass is pure white borosilicate glass of the first hydrolytic class.
  • Excellent chemical resistance
  • Optimal flatness
  • No disrupting fluorescence
Cover glass does not emit any disturbing fluorescence under the ultraviolet light of the fluorescence microscope.