Dual Small Hub Removable Needle Coupler, Hamilton Company
Supplier: Hamilton
55752-01
89187-004EA 95.52 USD
89187-004
Dual Small Hub Removable Needle Coupler, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes and accessories in the industry
The compression fittings and adaptors are designed to connect Hamilton small hub removable needle (RN) and knurled hub (KH) syringes to glass capillary tubing and PEEK tubing. The connectors can be combined in a variety of different configurations to meet most connection needs. Applications include neuroscience injections, patch clamp, cell transplantation, stem cell research, corrosive chemical dispensing, infusion pump injections and mass spectrometry iInfusion pumps. Luer to RN adaptor.
Each is manufactured to achieve the highest level of accuracy and precision possible.