Q700 Sonicator without a Probe, Qsonica
Supplier: QSonica, LLC.
Q700A-110
89184-808EA 8709.28 USD
89184-808
Q700 Sonicator without a Probe, Qsonica
Homogenizers Ultrasonic Cell Disruptors
Q700 Sonicator system without a probe
Probe/Cup Horn must be ordered separately.
Accessories information: Horn extenders, also available in solid or tapped varieties, can reach into long neck vessels. Half-wave extenders add 12.7 cm (5") to the horn’s length; full-wave extenders add 25.4 cm (10") to the horn’s length.
Delivery information: Supplied with power supply, converter, cables and wrench set.