Sonicator 700 Watts, Qsonica
Supplier: QSonica, LLC.
Q700-110
89184-804EA 9643.6 USD
89184-804
Sonicator 700 Watts, Qsonica
Homogenizers Ultrasonic Cell Disruptors
The new Q700 is the most technologically advanced Sonicator available today.
The Q700 is the only Sonicator on the market that offers full amplitude control from 1 - 100%. This enables greater control of the probe's intensity, helping to pinpoint the optimum settings for efficient sample processing. We have increased maximum power output to 700 watts making the system more durable and capable of handling even larger samples if necessary. Our new display, design improvements and added accessories make this the most sophisticated and versatile Sonicator available today.
A state-of-the-art touch screen interface offers intuitive control and provides a user-friendly experience. The most important feature of a Sonicator is reproducibility. Improved internal circuitry guarantees more efficient operation, sample-to-sample consistency and most importantly, a reliable end result.
Delivery information: Supplied with power supply, converter, cables, wrench set and standard ½” diameter probe with replaceable tip.