Corning® Polystyrene Roller Bottle with Easy Grip Cap
Supplier: Corning
431644
89184-640CS 1140.73 USD
89184-640
Corning® Polystyrene Roller Bottle with Easy Grip Cap
Flasks Cell Culture Flasks
The bottle is made with one piece seamless construction, has a wide mouth and printed graduations.
  • Orange easy grip cap features large knurls designed for ergonomic handling
  • Not tissue-culture treated to minimize cell retention during seeding
The bottle is manufactured from virgin polystyrene and the cap is made of high density polyethylene. Both resins meet the USP Class VI requirements for plastic containers and closures.
This product is sterilized by gamma radiation and is certified nonpyrogenic. It has printed lot numbers to aid in product traceability.
To be used with USP Class VI certified C-FLEX® Tubing Set (see Cat No. 89184-632).